(3-Bromo-2-chloro-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

98963-5G 98963-25G
77371-324EA 135.96 USD
77371-324 77383-330
(3-Bromo-2-chloro-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%
(3-Bromo-2-chloro-6-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃OS
Storage Temperature: Ambient
CAS Number: 2167830-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR