4-Bromo-2-(methoxymethyl)thiophene ≥97%

Supplier: AOBChem USA

Warning

98805-5G 98805-25G
77371-314EA 146.26 USD
77371-314 77383-320
4-Bromo-2-(methoxymethyl)thiophene ≥97%
4-Bromo-2-(methoxymethyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₇BrOS
MW: 207.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16095115
CAS Number: 141832-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR