N,N-Dimethyl-1H-pyrazole-1-sulfonamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N,N-Dimethyl-1H-pyrazole-1-sulfonamide

96725-5G 96725-25G
77371-280EA 70.04 USD
77371-280 77383-290
N,N-Dimethyl-1H-pyrazole-1-sulfonamide ≥97%
N,N-Dimethyl-1H-pyrazole-1-sulfonamide

Precaución: for Research Use Only

Formula: C₅H₉N₃O₂S
MW: 175.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11865226
CAS Number: 133228-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR