4-Ethoxy-1-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

95351-5G 95351-25G
77371-252EA 125.66 USD
77371-252 77383-264
4-Ethoxy-1-fluoro-2-methoxybenzene ≥97%
4-Ethoxy-1-fluoro-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774515
CAS Number: 2901099-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR