1-Bromo-2-(cyclopropylmethoxy)-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

89956-5G 89956-25G
77371-122EA 131.84 USD
77371-122 77383-130
1-Bromo-2-(cyclopropylmethoxy)-4-fluorobenzene ≥95%
1-Bromo-2-(cyclopropylmethoxy)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD24119184
CAS Number: 1173006-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR