Methyl 3-bromo-5-chloro-4-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

89465-5G 89465-25G
77371-098EA 163.77 USD
77371-098 77383-112
Methyl 3-bromo-5-chloro-4-hydroxybenzoate ≥97%
Methyl 3-bromo-5-chloro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
MW: 265.488 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25955178
CAS Number: 107356-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR