1,3-Dibromo-2-methoxy-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

87999-5G 87999-25G
77371-020EA 176.13 USD
77371-020 77383-060
1,3-Dibromo-2-methoxy-5-nitrobenzene ≥97%
1,3-Dibromo-2-methoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD00124312
CAS Number: 31106-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR