(4-Fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86075-5G 86075-25G
77370-902EA 83.43 USD
77370-902 77382-986
(4-Fluorophenyl)trimethylsilane ≥95%
(4-Fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₃FSi
MW: 168.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799543
CAS Number: 455-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR