2,4-Difluoro-6-iodoaniline ≥97%

Supplier: AOBChem USA

Warning

84880-5G 84880-25G
77370-828EA 70.04 USD
77370-828 77382-910
2,4-Difluoro-6-iodoaniline ≥97%
2,4-Difluoro-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
Storage Temperature: Ambient
MDL Number: MFCD08059536
CAS Number: 582319-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR