3-Fluoro-4-methoxybenzamide ≥95%

Supplier: AOBChem USA

Warning

84173-5G 84173-25G
77370-804EA 149.35 USD
77370-804 77382-888
3-Fluoro-4-methoxybenzamide ≥95%
3-Fluoro-4-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660163
CAS Number: 701640-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR