(3-Iodo-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

83434-5G 83434-25G
77370-780EA 174.07 USD
77370-780 77382-862
(3-Iodo-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(3-Iodo-2-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁IO₂S
MW: 310.152 g/mol
Storage Temperature: Ambient
CAS Number: 197149-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR