Ethyl-2,4-dichlorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2,4-dichlorobenzoylformate

82403-5G 82403-25G
77370-664EA 67.98 USD
77370-664 77382-800
Ethyl-2,4-dichlorobenzoylformate ≥95%
Ethyl-2,4-dichlorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094603
CAS Number: 34966-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR