Ethyl-3,4-difluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82394-5G 82394-25G
77370-660EA 103 USD
77370-660 77382-796
Ethyl-3,4-difluorobenzoylformate ≥95%
Ethyl 3,4-difluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01319619
CAS Number: 73790-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR