Ethyl-3,4-dichlorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82390-5G 82390-25G
77370-656EA 64.89 USD
77370-656 77382-792
Ethyl-3,4-dichlorobenzoylformate ≥95%
Ethyl 3,4-dichlorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01934864
CAS Number: 34966-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR