2-Iodo-1,3,4-trimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

82277-5G 82277-25G
77370-638EA 173.04 USD
77370-638 77382-776
2-Iodo-1,3,4-trimethoxybenzene ≥97%
2-Iodo-1,3,4-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₃
MW: 294.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769255
CAS Number: 25245-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR