N,4-Dimethoxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

81878-5G 81878-25G
77370-624EA 83.43 USD
77370-624 77382-758
N,4-Dimethoxy-N-methylbenzamide ≥95%
N,4-Dimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD02684314
CAS Number: 52898-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR