1-Bromo-5-chloro-2-nitro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

81835-5G 81835-25G
77370-622EA 90.64 USD
77370-622 77382-756
1-Bromo-5-chloro-2-nitro-4-(trifluoromethyl)benzene ≥97%
1-Bromo-5-chloro-2-nitro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrClF₃NO₂
MW: 304.448 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768990
CAS Number: 2169477-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR