1-Fluoro-2-methoxy-4-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

81412-5G 81412-25G
77370-596EA 172.01 USD
77370-596 77382-734
1-Fluoro-2-methoxy-4-(methoxymethyl)benzene ≥97%
1-Fluoro-2-methoxy-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768790
CAS Number: 2969242-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR