Ethyl-4-methoxybenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-methoxybenzoylformate

81296-5G 81296-25G
77370-588EA 60.77 USD
77370-588 77382-718
Ethyl-4-methoxybenzoylformate ≥97%
Ethyl 4-methoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
MW: 208.2106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00963513
CAS Number: 40140-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR