4,5-Dibromo-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

80558-5G 80558-25G
77370-540EA 131.84 USD
77370-540 77382-666
4,5-Dibromo-2-fluorophenol ≥97%
4,5-Dibromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD11870650
CAS Number: 1155354-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR