1,3-Dibromo-5-butylbenzene ≥95%

Supplier: AOBChem USA

Warning

80530-5G 80530-25G
77370-538EA 152.44 USD
77370-538 77382-664
1,3-Dibromo-5-butylbenzene ≥95%
1,3-Dibromo-5-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂
Storage Temperature: Ambient
MDL Number: MFCD11845939
CAS Number: 75894-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR