3,4-Difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

80462-5G 80462-25G
77370-522EA 100.94 USD
77370-522 77382-648
3,4-Difluoro-N-methoxy-N-methylbenzamide ≥97%
3,4-Difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684299
CAS Number: 188345-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR