(2-Fluoro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

79918-5G 79918-25G
77370-484EA 131.84 USD
77370-484 77382-600
(2-Fluoro-4-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₉FS
Storage Temperature: Ambient
MDL Number: MFCD15527257
CAS Number: 1378582-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR