(3-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

79913-5G 79913-25G
77370-478EA 83.43 USD
77370-478 77382-598
(3-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane ≥97%
(3-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD34768208
CAS Number: 2706348-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR