2-Bromo-4-isopropoxy-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

79708-5G 79708-25G
77370-456EA 131.84 USD
77370-456 77382-568
2-Bromo-4-isopropoxy-1-methoxybenzene ≥97%
2-Bromo-4-isopropoxy-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356579
CAS Number: 124764-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR