2,6-Dichloro-N-cyclohexylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

77416-5G 77416-25G
77370-320EA 181.28 USD
77370-320 77382-418
2,6-Dichloro-N-cyclohexylnicotinamide ≥97%
2,6-Dichloro-N-cyclohexylnicotinamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD23132156
CAS Number: 1003872-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR