2,6-Bis(benzyloxy)-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

77110-5G 77110-25G
77370-304EA 138.02 USD
77370-304 77382-382
2,6-Bis(benzyloxy)-3-iodopyridine ≥97%
2,6-Bis(benzyloxy)-3-iodopyridine

Precaución: for Research Use Only

Formula: C₁₉H₁₆INO₂
MW: 417.2403 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34706499
CAS Number: 2259853-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR