4-Bromo-1-isobutoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

76797-5G 76797-25G
77370-270EA 100.94 USD
77370-270 77382-336
4-Bromo-1-isobutoxy-2-(trifluoromethyl)benzene ≥97%
4-Bromo-1-isobutoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₃O
MW: 297.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27931152
CAS Number: 876918-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR