5-(Benzyloxy)-2-bromo-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

76747-5G 76747-25G
77370-264EA 183.34 USD
77370-264 77382-332
5-(Benzyloxy)-2-bromo-3-methylpyridine ≥97%
5-(Benzyloxy)-2-bromo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD34766698
CAS Number: 2901098-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR