tert-Butyl 3-chloro-2,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

75885-5G 75885-25G
77370-178EA 131.84 USD
77370-178 77382-256
tert-Butyl 3-chloro-2,6-difluorobenzoate ≥97%
tert-Butyl 3-chloro-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClF₂O₂
MW: 248.6536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766201
CAS Number: 2901107-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR