(2,6-Difluoro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

75421-5G 75421-25G
77370-132EA 147.29 USD
77370-132 77382-210
(2,6-Difluoro-4-methylphenyl)(methyl)sulfane ≥97%
(2,6-Difluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂S
MW: 174.2109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790542
CAS Number: 1803808-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR