3,4',5-Tribromo-[1,1'-biphenyl]-4-ol ≥95%

Supplier: AOBChem USA

Warning

74464-5G 74464-25G
77370-026EA 91.67 USD
77370-026 77382-102
3,4',5-Tribromo-[1,1'-biphenyl]-4-ol ≥95%
3,4',5-Tribromo-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₇Br₃O
MW: 406.895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765488
CAS Number: 4544-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR