4-Chloro-6-methoxynicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

72916-5G 72916-25G
77369-868EA 170.98 USD
77369-868 77381-934
4-Chloro-6-methoxynicotinaldehyde ≥95%
4-Chloro-6-methoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13188719
CAS Number: 1060806-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR