Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

72836-5G 72836-25G
77369-858EA 149.35 USD
77369-858 77381-924
Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate ≥97%
Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17206126
CAS Number: 139308-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR