1-(tert-Butyl)-4-(methylthio)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

72833-5G 72833-25G
77369-856EA 123.6 USD
77369-856 77381-922
1-(tert-Butyl)-4-(methylthio)-1H-pyrazole ≥97%
1-(tert-Butyl)-4-(methylthio)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₄N₂S
MW: 170.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33180180
CAS Number: 2805968-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR