1-Ethoxy-2-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72786-5G 72786-25G
77369-836EA 173.04 USD
77369-836 77381-898
1-Ethoxy-2-fluoro-4-methoxybenzene ≥97%
1-Ethoxy-2-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764552
CAS Number: 1026793-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR