1,2-Dibromo-5-chloro-3-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72587-5G 72587-25G
77369-772EA 62.83 USD
77369-772 77381-838
1,2-Dibromo-5-chloro-3-ethoxybenzene ≥97%
1,2-Dibromo-5-chloro-3-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂ClO
MW: 314.402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764434
CAS Number: 2387151-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR