1,2-Dibromo-5-chloro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72586-5G 72586-25G
77369-770EA 60.77 USD
77369-770 77381-836
1,2-Dibromo-5-chloro-3-isopropoxybenzene ≥97%
1,2-Dibromo-5-chloro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Br₂ClO
MW: 328.428 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764433
CAS Number: 2901082-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR