1-Ethoxy-3,4-difluoro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

71380-5G 71380-25G
77369-608EA 64.89 USD
77369-608 77381-682
1-Ethoxy-3,4-difluoro-2-iodobenzene ≥95%
1-Ethoxy-3,4-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
MW: 284.0418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764111
CAS Number: 2384732-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR