1,2-Diiodo-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

69076-5G 69076-25G
77369-428EA 60.77 USD
77369-428 77381-516
1,2-Diiodo-4-fluorobenzene ≥95%
1,2-Diiodo-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃FI₂
Storage Temperature: Ambient
MDL Number: MFCD00042576
CAS Number: 203059-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR