Trimethyl(o-tolyl)silane ≥97%

Supplier: AOBChem USA

Warning

68830-5G 68830-25G
77369-334EA 131.84 USD
77369-334 77381-430
Trimethyl(o-tolyl)silane ≥97%
Trimethyl(o-tolyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₆Si
MW: 164.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094130
CAS Number: 7450-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR