1-Bromo-4-isobutoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

68573-5G 68573-25G
77369-284EA 100.94 USD
77369-284 77381-392
1-Bromo-4-isobutoxy-2-methylbenzene ≥95%
1-Bromo-4-isobutoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917739
CAS Number: 1309932-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR