4'-Ethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

67768-5G 67768-25G
77369-034EA 170.98 USD
77369-034 77381-132
4'-Ethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%
4'-Ethoxy-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799358
CAS Number: 729-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR