5-Bromo-2-chloro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66235-5G 66235-25G
77368-812EA 133.9 USD
77368-812 77380-938
5-Bromo-2-chloro-1,1'-biphenyl ≥97%
5-Bromo-2-chloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrCl
MW: 267.549 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30098067
CAS Number: 126866-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR