1-Bromo-4-((4-Bromobenzyl)oxy)-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

65530-5G 65530-25G
77368-624EA 94.76 USD
77368-624 77380-834
1-Bromo-4-((4-Bromobenzyl)oxy)-2-methoxybenzene ≥97%
1-Bromo-4-((4-Bromobenzyl)oxy)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂O₂
MW: 372.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762249
CAS Number: 2807456-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR