Thiomorpholino(2,3,6-trifluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

65429-5G 65429-25G
77368-576EA 142.14 USD
77368-576 77380-790
Thiomorpholino(2,3,6-trifluorophenyl)methanone ≥97%
Thiomorpholino(2,3,6-trifluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₃NOS
Storage Temperature: Ambient
MDL Number: MFCD34762211
CAS Number: 2807440-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR