2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloropyridin-3-yl)(pyrrolidin-1-yl)methanone

64845-5G 64845-25G
77368-414EA 123.6 USD
77368-414 77380-612
2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%
2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O
MW: 210.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01342311
CAS Number: 60597-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR