3-Chloro-5-fluoro-N,N-dimethylaniline ≥95%

Supplier: AOBChem USA

Warning

64839-5G 64839-25G
77368-408EA 119.48 USD
77368-408 77380-606
3-Chloro-5-fluoro-N,N-dimethylaniline ≥95%
3-Chloro-5-fluoro-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₉ClFN
MW: 173.6151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091137
CAS Number: 1369918-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR