1,3-Dibromo-2-fluoro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

64619-5G 64619-25G
77368-330EA 166.86 USD
77368-330 77380-550
1,3-Dibromo-2-fluoro-5-(trifluoromethyl)benzene ≥97%
1,3-Dibromo-2-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₂F₄
MW: 321.8923 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784692
CAS Number: 109919-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR