Methyl 2,3-difluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

64389-5G 64389-25G
77368-256EA 145.23 USD
77368-256 77380-488
Methyl 2,3-difluoroisonicotinate ≥97%
Methyl 2,3-difluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21090433
CAS Number: 1353102-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR